Assay Details
| Gene Symbol: LOC100409586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746172.3 | XP_002746218.2 | 3 - 4 | 816 | 104 | |
| XM_008993844.2 | XP_008992092.1 | 3 - 4 | 736 | 104 | ||
| XM_008993845.2 | XP_008992093.1 | 3 - 4 | 913 | 104 | ||
| XM_008993846.2 | XP_008992094.1 | 3 - 4 | 913 | 104 | ||
| XM_008993847.2 | XP_008992095.1 | 3 - 4 | 913 | 104 | ||
| XM_008993848.2 | XP_008992096.1 | 3 - 4 | 913 | 104 | ||
More Information
Set Membership: |
|

LOC100409586
Genomic Map