Assay Details
| Gene Symbol: LOC100409586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993847.2 | XP_008992095.1 | 6 - 7 | 1436 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100409586| Gene Symbol: LOC100409586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993847.2 | XP_008992095.1 | 6 - 7 | 1436 | 102 | |
Set Membership: |
|