Assay Details
| Gene Symbol: LOC100415296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981861.2 | XP_008980109.2 | 46 - 47 | 7839 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100415296| Gene Symbol: LOC100415296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981861.2 | XP_008980109.2 | 46 - 47 | 7839 | 88 | |
Set Membership: |
|