Assay Details
| Gene Symbol: LOC100411501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746372.4 | XP_002746418.3 | 25 - 26 | 3309 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100411501| Gene Symbol: LOC100411501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746372.4 | XP_002746418.3 | 25 - 26 | 3309 | 90 | |
Set Membership: |
|