Assay Details
| Gene Symbol: MYPOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807853.4 | XP_002807899.2 | 2 - 3 | 605 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYPOP| Gene Symbol: MYPOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807853.4 | XP_002807899.2 | 2 - 3 | 605 | 109 | |
Set Membership: |
|