Assay Details
| Gene Symbol: CYP19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267762.1 | NP_001254691.1 | 3 - 4 | 451 | 64 | |
| XM_009005416.2 | XP_009003664.1 | 4 - 5 | 558 | 64 | ||
| XM_009005417.2 | XP_009003665.1 | 4 - 5 | 1327 | 64 | ||
| XM_009005419.2 | XP_009003667.1 | 4 - 5 | 507 | 64 | ||
| GenBank mRNA | AY034779.2 | - | 3 - 4 | 451 | 64 | |
More Information
Set Membership: |
|

CYP19A1
Genomic Map