Assay Details
| Gene Symbol: B3GALT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971017.1 | XP_017826506.1 | 1 | 1415 | 71 | |
| GenBank mRNA | EF215243.1 | - | 1 | 419 | 71 | |
| EF215355.1 | - | 1 | 419 | 71 | ||
More Information
Set Membership: |
|

B3GALT4
Genomic Map