Assay Details
| Gene Symbol: TCP11L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007890.2 | XP_009006138.1 | 7 - 8 | 1366 | 58 | |
| XM_009007891.2 | XP_009006139.1 | 7 - 8 | 1385 | 58 | ||
| XM_017977997.1 | XP_017833486.1 | 7 - 8 | 1385 | 58 | ||
More Information
Set Membership: |
|

TCP11L1
Genomic Map