Assay Details
| Gene Symbol: NFYA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746523.4 | XP_002746569.1 | 4 - 5 | 477 | 94 | |
| XM_008994389.2 | XP_008992637.1 | 4 - 5 | 487 | 94 | ||
| XM_008994391.2 | XP_008992639.1 | 4 - 5 | 472 | 94 | ||
| XM_008994392.2 | XP_008992640.1 | 4 - 5 | 484 | 94 | ||
| XM_008994393.2 | XP_008992641.1 | 4 - 5 | 484 | 94 | ||
| XM_008994395.2 | XP_008992643.1 | 4 - 5 | 487 | 94 | ||
| XM_008994396.2 | XP_008992644.1 | 3 - 4 | 1649 | 94 | ||
| XM_008994397.2 | XP_008992645.1 | 4 - 5 | 585 | 94 | ||
| XM_008994400.2 | XP_008992648.1 | 3 - 4 | 1649 | 94 | ||
| XM_017971066.1 | XP_017826555.1 | 4 - 5 | 487 | 94 | ||
| XM_017971067.1 | XP_017826556.1 | 4 - 5 | 487 | 94 | ||
More Information
Set Membership: |
|

NFYA
Genomic Map