Assay Details
| Gene Symbol: MSMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992496.2 | XP_008990744.1 | 6 - 7 | 949 | 95 | |
| XM_017970162.1 | XP_017825651.1 | 5 - 6 | 655 | 95 | ||
More Information
Set Membership: |
|

MSMO1
Genomic Map