Assay Details
| Gene Symbol: DGUOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757603.4 | XP_002757649.1 | 6 - 7 | 884 | 89 | |
| XM_008980493.2 | XP_008978741.1 | 4 - 5 | 643 | 89 | ||
| XM_017964233.1 | XP_017819722.1 | 5 - 6 | 782 | 89 | ||
| XM_017964234.1 | XP_017819723.1 | 5 - 6 | 572 | 89 | ||
More Information
Set Membership: |
|

DGUOK
Genomic Map