Assay Details
| Gene Symbol: PIR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762645.3 | XP_002762691.1 | 5 - 6 | 754 | 75 | |
| XM_008988989.2 | XP_008987237.1 | 5 - 6 | 762 | 75 | ||
| XM_017968478.1 | XP_017823967.1 | 5 - 6 | 1004 | 75 | ||
More Information
Set Membership: |
|

PIR
Genomic Map