Assay Details
| Gene Symbol: CENPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761193.4 | XP_002761239.1 | 10 - 11 | 1205 | 71 | |
| XM_017966980.1 | XP_017822469.1 | 10 - 11 | 1022 | 71 | ||
| XM_017966981.1 | XP_017822470.1 | 9 - 10 | 1115 | 71 | ||
| XM_017966982.1 | XP_017822471.1 | 9 - 10 | 1103 | 71 | ||
More Information
Set Membership: |
|

CENPN
Genomic Map