Assay Details
| Gene Symbol: LOC100388660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970726.1 | XP_017826215.1 | 10 - 11 | 1028 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100388660| Gene Symbol: LOC100388660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970726.1 | XP_017826215.1 | 10 - 11 | 1028 | 107 | |
Set Membership: |
|