Assay Details
| Gene Symbol: LOC100388660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970726.1 | XP_017826215.1 | 15 - 16 | 1683 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100388660| Gene Symbol: LOC100388660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970726.1 | XP_017826215.1 | 15 - 16 | 1683 | 58 | |
Set Membership: |
|