Assay Details
| Gene Symbol: LOC100387213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760499.4 | XP_002760545.1 | 17 - 18 | 2280 | 62 | |
| XM_008985188.2 | XP_008983436.1 | 17 - 18 | 2280 | 62 | ||
More Information
Set Membership: |
|

LOC100387213
Genomic Map