Assay Details
| Gene Symbol: EGFL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743465.3 | XP_002743511.1 | 8 - 9 | 2380 | 87 | |
| XM_009005532.1 | XP_009003780.1 | 7 - 8 | 810 | 87 | ||
| XM_009005536.1 | XP_009003784.1 | 7 - 8 | 765 | 87 | ||
| XM_009005542.2 | XP_009003790.1 | 9 - 10 | 1109 | 87 | ||
| XM_009005546.1 | XP_009003794.1 | 8 - 9 | 880 | 87 | ||
| XM_009005553.1 | XP_009003801.1 | 7 - 8 | 699 | 87 | ||
More Information
Set Membership: |
|

EGFL7
Genomic Map