Assay Details
| Gene Symbol: NARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754768.4 | XP_002754814.1 | 6 - 7 | 1048 | 88 | |
| XM_009007359.2 | XP_009005607.1 | 6 - 7 | 1025 | 88 | ||
| XM_009007361.2 | XP_009005609.1 | 6 - 7 | 1026 | 88 | ||
| XM_009007363.2 | XP_009005611.1 | 6 - 7 | 573 | 88 | ||
| XM_017977757.1 | XP_017833246.1 | 6 - 7 | 1028 | 88 | ||
| XM_017977759.1 | XP_017833248.1 | 6 - 7 | 1028 | 88 | ||
| XM_017977760.1 | XP_017833249.1 | 6 - 7 | 569 | 88 | ||
| XM_017977761.1 | XP_017833250.1 | 5 - 6 | 460 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009007363.2, XM_017977760.1, XM_017977761.1 transcripts.
Set Membership: |
|

NARS2
Genomic Map