Assay Details
| Gene Symbol: KCNS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759363.3 | XP_002759409.1 | 1 - 2 | 395 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002759363.3 transcripts.
Set Membership: |
|

KCNS2
Genomic Map