Assay Details
| Gene Symbol: LOC100412883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008889.2 | XP_009007137.2 | 5 - 6 | 786 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100412883| Gene Symbol: LOC100412883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008889.2 | XP_009007137.2 | 5 - 6 | 786 | 73 | |