Assay Details
| Gene Symbol: DBF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002527.1 | XP_009000775.1 | 7 - 8 | 931 | 87 | |
| XM_017975384.1 | XP_017830873.1 | 7 - 8 | 1130 | 87 | ||
| XM_017975385.1 | XP_017830874.1 | 5 - 6 | 240 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017975385.1 transcripts.
Set Membership: |
|

DBF4
Genomic Map