Assay Details
| Gene Symbol: P2RX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747849.2 | XP_002747895.1 | 12 - 13 | 1408 | 111 | |
| XM_008996406.1 | XP_008994654.1 | 12 - 13 | 1407 | 111 | ||
| XM_008996407.1 | XP_008994655.1 | 11 - 12 | 1326 | 111 | ||
More Information
Set Membership: |
|

P2RX1
Genomic Map