Assay Details
| Gene Symbol: MRPL52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753739.4 | XP_002753785.3 | 3 - 4 | 291 | 71 | |
| XM_009005736.2 | XP_009003984.1 | 3 - 4 | 294 | 71 | ||
More Information
Set Membership: |
|

MRPL52
Genomic Map