Assay Details
| Gene Symbol: ANXA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992477.1 | XP_008990725.1 | 6 - 7 | 666 | 84 | |
| XM_017970158.1 | XP_017825647.1 | 4 - 5 | 409 | 84 | ||
| XM_017970159.1 | XP_017825648.1 | 4 - 5 | 380 | 84 | ||
More Information
Set Membership: |
|

ANXA10
Genomic Map