Assay Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754187.3 | XP_002754233.1 | 2 - 3 | 597 | 103 | |
| XM_009006431.2 | XP_009004679.1 | 2 - 3 | 597 | 103 | ||
| XM_017977284.1 | XP_017832773.1 | 3 - 4 | 799 | 103 | ||
| XM_017977285.1 | XP_017832774.1 | 3 - 4 | 2698 | 103 | ||
| XM_017977286.1 | XP_017832775.1 | 3 - 4 | 785 | 103 | ||
| XM_017977287.1 | XP_017832776.1 | 3 - 4 | 799 | 103 | ||
| XM_017977288.1 | XP_017832777.1 | 3 - 4 | 799 | 103 | ||
More Information
Set Membership: |
|

FOXN3
Genomic Map