Assay Details
| Gene Symbol: RBM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754426.2 | XP_002754472.1 | 2 - 3 | 398 | 78 | |
| XM_009006835.1 | XP_009005083.1 | 2 - 3 | 398 | 78 | ||
More Information
Set Membership: |
|

RBM7
Genomic Map