Assay Details
| Gene Symbol: THNSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980361.2 | XP_008978609.1 | 4 - 5 | 674 | 62 | |
| XM_008980362.2 | XP_008978610.1 | 4 - 5 | 677 | 62 | ||
| XM_008980363.2 | XP_008978611.1 | 3 - 4 | 2914 | 62 | ||
| XM_017964180.1 | XP_017819669.1 | 4 - 5 | 677 | 62 | ||
More Information
Set Membership: |
|

THNSL2
Genomic Map