Assay Details
| Gene Symbol: RPLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755680.4 | XP_002755726.1 | 2 - 3 | 241 | 77 | |
| XM_009008752.2 | XP_009007000.1 | 2 - 3 | 251 | 77 | ||
More Information
Set Membership: |
|

RPLP2
Genomic Map