Assay Details
| Gene Symbol: KLF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750002.2 | XP_002750048.1 | 1 - 2 | 371 | 56 | |
| XM_008999852.1 | XP_008998100.1 | 1 - 2 | 371 | 56 | ||
More Information
Set Membership: |
|

KLF6
Genomic Map