Assay Details
| Gene Symbol: CYB5D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747851.3 | XP_002747897.2 | 2 - 3 | 872 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYB5D2| Gene Symbol: CYB5D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747851.3 | XP_002747897.2 | 2 - 3 | 872 | 90 | |
Set Membership: |
|