Assay Details
| Gene Symbol: MRPS34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755760.2 | XP_002755806.1 | 2 - 3 | 434 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPS34| Gene Symbol: MRPS34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755760.2 | XP_002755806.1 | 2 - 3 | 434 | 100 | |
Set Membership: |
|