Assay Details
Gene Symbol: TMEM218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009007054.2 | XP_009005302.1 | 3 - 4 | 538 | 79 | |
XM_009007055.2 | XP_009005303.1 | 3 - 4 | 506 | 79 | ||
XM_009007056.2 | XP_009005304.1 | 4 - 5 | 532 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
