Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995701.2 | XP_008993949.1 | 2 - 3 | 791 | 63 | |
| XM_017971939.1 | XP_017827428.1 | 3 - 4 | 919 | 63 | ||
| GenBank mRNA | KC415025.1 | - | 1 - 2 | 119 | 63 | |
More Information
Set Membership: |
|

MAVS
Genomic Map