Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753503.3 | XP_002753549.1 | 5 - 6 | 1032 | 70 | |
| XM_002753504.3 | XP_002753550.1 | 5 - 6 | 1037 | 70 | ||
| XM_009005483.2 | XP_009003731.1 | 5 - 6 | 785 | 70 | ||
More Information
Set Membership: |
|

DNAAF4
Genomic Map