Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753503.3 | XP_002753549.1 | 6 - 7 | 1135 | 76 | |
| XM_002753504.3 | XP_002753550.1 | 6 - 7 | 1140 | 76 | ||
| XM_009005483.2 | XP_009003731.1 | 6 - 7 | 888 | 76 | ||
More Information
Set Membership: |
|

DNAAF4
Genomic Map