Assay Details
| Gene Symbol: ATP6AP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991865.2 | XP_008990113.1 | 5 - 6 | 1198 | 85 | |
| XM_017969805.1 | XP_017825294.1 | 5 - 6 | 1207 | 85 | ||
| XM_017969806.1 | XP_017825295.1 | 6 - 7 | 1320 | 85 | ||
| XM_017969807.1 | XP_017825296.1 | 5 - 6 | 503 | 85 | ||
| XM_017969808.1 | XP_017825297.1 | 5 - 6 | 1274 | 85 | ||
| XM_017969809.1 | XP_017825298.1 | 7 - 8 | 1425 | 85 | ||
| XM_017969810.1 | XP_017825299.1 | 5 - 6 | 1057 | 85 | ||
| XM_017969811.1 | XP_017825300.1 | 2 - 3 | 343 | 85 | ||
More Information
Set Membership: |
|

ATP6AP1L
Genomic Map