Assay Details
| Gene Symbol: PITX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745478.4 | XP_002745524.1 | 2 - 3 | 1061 | 60 | |
| XM_002745479.4 | XP_002745525.1 | 4 - 5 | 1831 | 60 | ||
| XM_008992842.2 | XP_008991090.1 | 5 - 6 | 1969 | 60 | ||
More Information
Set Membership: |
|

PITX2
Genomic Map