Assay Details
| Gene Symbol: F11R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984758.2 | XP_008983006.1 | 1 - 2 | 339 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F11R| Gene Symbol: F11R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984758.2 | XP_008983006.1 | 1 - 2 | 339 | 58 | |
Set Membership: |
|