Assay Details
| Gene Symbol: DDT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743606.4 | XP_002743652.1 | 1 - 2 | 277 | 52 | |
| XM_009008048.2 | XP_009006296.1 | 1 - 2 | 197 | 52 | ||
More Information
Set Membership: |
|

DDT
Genomic Map