Assay Details
| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963690.1 | XP_017819179.1 | 9 | 2701 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF10D| Gene Symbol: TNFRSF10D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963690.1 | XP_017819179.1 | 9 | 2701 | 72 | |
Set Membership: |
|