Assay Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008980004.2 | XP_008978252.2 | 2 - 3 | 2081 | 94 | |
XM_017963561.1 | XP_017819050.1 | 2 - 3 | 3078 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
