Assay Details
Gene Symbol: EGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753799.4 | XP_002753845.1 | 3 - 4 | 944 | 69 | |
XM_017977046.1 | XP_017832535.1 | 3 - 4 | 660 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
