Assay Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745287.3 | XP_002745333.1 | 10 - 11 | 1247 | 65 | |
| XM_008992540.2 | XP_008990788.1 | 11 - 12 | 1415 | 65 | ||
| XM_008992542.2 | XP_008990790.1 | 9 - 10 | 963 | 65 | ||
More Information
Set Membership: |
|

TMEM144
Genomic Map