Assay Details
| Gene Symbol: TXNL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761119.4 | XP_002761165.1 | 3 - 4 | 538 | 84 | |
| XM_017966893.1 | XP_017822382.1 | 3 - 4 | 491 | 84 | ||
More Information
Set Membership: |
|

TXNL4B
Genomic Map