Assay Details
| Gene Symbol: C21H3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735367.3 | XP_003735415.1 | 2 - 3 | 698 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C21H3orf38| Gene Symbol: C21H3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735367.3 | XP_003735415.1 | 2 - 3 | 698 | 88 | |
Set Membership: |
|