Assay Details
| Gene Symbol: LOC100409865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974322.1 | XP_017829811.1 | 20 - 21 | 4044 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100409865| Gene Symbol: LOC100409865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974322.1 | XP_017829811.1 | 20 - 21 | 4044 | 80 | |
Set Membership: |
|