Assay Details
| Gene Symbol: CYR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749848.3 | XP_002749894.2 | 4 - 5 | 1092 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYR61| Gene Symbol: CYR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749848.3 | XP_002749894.2 | 4 - 5 | 1092 | 68 | |
Set Membership: |
|