Assay Details
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009009671.2 | XP_009007919.1 | 7 - 8 | 1309 | 70 | |
XM_009009672.2 | XP_009007920.2 | 7 - 8 | 1210 | 70 | ||
XM_009009673.2 | XP_009007921.1 | 6 - 7 | 1119 | 70 | ||
XM_009009675.2 | XP_009007923.1 | 3 - 4 | 758 | 70 | ||
XM_009009677.2 | XP_009007925.1 | 3 - 4 | 1427 | 70 | ||
XM_017978977.1 | XP_017834466.1 | 4 - 5 | 799 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
