Assay Details
| Gene Symbol: CDIPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756048.4 | XP_002756094.1 | 4 - 5 | 827 | 70 | |
| XM_017978855.1 | XP_017834344.1 | 4 - 5 | 710 | 70 | ||
| XM_017978856.1 | XP_017834345.1 | 4 - 5 | 366 | 70 | ||
More Information
Set Membership: |
|

CDIPT
Genomic Map