Assay Details
| Gene Symbol: DCTD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992414.2 | XP_008990662.1 | 3 - 4 | 419 | 90 | |
| XM_008992415.1 | XP_008990663.1 | 3 - 4 | 705 | 90 | ||
| XM_008992416.2 | XP_008990664.1 | 3 - 4 | 308 | 90 | ||
| XM_008992417.2 | XP_008990665.1 | 3 - 4 | 476 | 90 | ||
More Information
Set Membership: |
|

DCTD
Genomic Map